Learn More
Cayman Chemical ClemIzolehydrochlorIde 50mg
SDP
Catalog No. 502967249
Encompass_Preferred
An antihistamine that antagonizes the H1 receptor at high nanomolar concentrations; less potently blocks TRPC5 (IC50 = 1.0-1.3 µM), with at least 6-fold selectivity for TRPC5 over other TRP channels
Catalog No. 50-296-7249 Supplier Cayman Chemical Supplier No. 1769550 MG
An antihistamine that antagonizes the H1 receptor at high nanomolar concentrations; less potently blocks TRPC5 (IC50 = 1.0-1.3 µM), with at least 6-fold selectivity for TRPC5 over other TRP channels