Learn More
Cayman Chemical Clenbuterol-d9, Purity: ≥99% deuterated forms (d1-d9), CAS: 129138-58-5, 10 mg
SDP
Catalog No. 502942583
Encompass_Preferred
An internal standard for the quantification of clenbuterol by GC- or LC-MS
Catalog No. 50-294-2583 Supplier Cayman Chemical Supplier No. 4081610 MG
An internal standard for the quantification of clenbuterol by GC- or LC-MS