Learn More
Medchemexpress LLC Clenbuterol-d9 | 129138-58-5 | 99.9% | 1 MG
SDP
Catalog No. 5000412718
Encompass_Preferred
Clenbuterol-d9 is the deuterium labeled Clenbuterol, acting as a β2-adrenergic receptor agonist. This compound is suitable for use as a tracer and an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • β2-adrenergic receptor agonist
  • Useful as a tracer in analytical techniques
  • Functions as an internal standard for quantitative analysis
Catalog No. 50-004-12718 Supplier Medchemexpress LLC Supplier No. HYB1615S1MG
Clenbuterol-d9 is the deuterium labeled Clenbuterol, acting as a β2-adrenergic receptor agonist. This compound is suitable for use as a tracer and an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • β2-adrenergic receptor agonist
  • Useful as a tracer in analytical techniques
  • Functions as an internal standard for quantitative analysis