Learn More
Abcam Clenbuterol ELISA Kit, 1 x 96 Tests
Catalog No. 502692038
Encompass_Preferred
Clenbuterol ELISA Kit is a Competitive ELISA kit for the measurement of Clenbuterol in Food samples, Tissue, Urine samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-2038 Supplier Abcam Supplier No. AB3029361X96TESTS
Clenbuterol ELISA Kit is a Competitive ELISA kit for the measurement of Clenbuterol in Food samples, Tissue, Urine samples.
The product is subject to the following: Abcam Restricted Use Statement