Learn More
Medchemexpress LLC Clenbuterol hydrochloride | 21898-19-1 | 99.9% | 10 MM 1 ML
SDP
Catalog No. 5000411096
Encompass_Preferred
Clenbuterol hydrochloride is a selective β2-adrenergic agonist that enhances skeletal muscle strength and hypertrophy. It induces growth factor mRNA, activates astrocytes, and protects rat brain tissue against ischemic damage.
  • Enhances skeletal muscle strength and hypertrophy.
  • Induces growth factor mRNA.
  • Activates astrocytes.
  • Protects rat brain tissue against ischemic damage.
Catalog No. 50-004-11096 Supplier Medchemexpress LLC Supplier No. HYB161410MM/1ML
Clenbuterol hydrochloride is a selective β2-adrenergic agonist that enhances skeletal muscle strength and hypertrophy. It induces growth factor mRNA, activates astrocytes, and protects rat brain tissue against ischemic damage.
  • Enhances skeletal muscle strength and hypertrophy.
  • Induces growth factor mRNA.
  • Activates astrocytes.
  • Protects rat brain tissue against ischemic damage.