Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab, Inc. Clerical Medical Labels

Catalog No. 22505490
Change view
Click to view available options
Length (English):
0.375 in.
0.563 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-505-490 0.375 in.
22-505-463 0.563 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-505-490 Supplier Marketlab, Inc. Supplier No. 9097
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A convenient resource for Clerical Medical Label messaging needs.

  • Blister packaging for convenient product handling
  • Includes Clerical Medical Labels for a variety of messaging needs
  • STAT labels are available for urgent labeling requirements
  • PRIORITY labels support priority messaging needs
  • TLC labels are available for specific clerical medical labeling requirements

Specifications

Adhesive Type Peel and Stick
Color Orange
Finish Opaque
Format Label
Material Paper
Product Type Label
Sterility Non-sterile
Length (English) 0.375 in.
Width (English) 1.625 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.