Learn More
Apexbio Technology LLC Clevidipine Butyrate 167221-71-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092321
Encompass_Preferred
Clevidipine butyrate (CAS 167221-71-8) is a rapidly acting dihydropyridine calcium channel antagonist that selectively inhibits L-type calcium channels in vascular smooth muscle This inhibition leads to arterial vasodilation and subsequent reduction in blood pressure In vitro studies demonstrate an IC50 of 7 1 nM at a holding potential of -40 mV indicating high potency for its target Clevidipine butyrate is primarily investigated in the context of perioperative hypertension offering a research tool for evaluating acute blood pressure management and the pharmacodynamics of short-acting calcium channel blockade in vascular systems
Catalog No. 50-000-92321 Supplier Apexbio Technology LLC Supplier No. B14155.1
Clevidipine butyrate (CAS 167221-71-8) is a rapidly acting dihydropyridine calcium channel antagonist that selectively inhibits L-type calcium channels in vascular smooth muscle This inhibition leads to arterial vasodilation and subsequent reduction in blood pressure In vitro studies demonstrate an IC50 of 7 1 nM at a holding potential of -40 mV indicating high potency for its target Clevidipine butyrate is primarily investigated in the context of perioperative hypertension offering a research tool for evaluating acute blood pressure management and the pharmacodynamics of short-acting calcium channel blockade in vascular systems