Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clevidipine butyrate impurity 1 | 123853-39-4 | 95.1% | 356.20 | 5 G
SDP

Catalog No. 5000401667
Encompass_Preferred

Clevidipine butyrate impurity 1 is an impurity of Clevidipine butyrate. It is for research use only and not intended for human use. The product is sold to researchers and not to patients.

  • Appearance: Off-white to light yellow solid
  • Formula: C16H15Cl2NO4
  • SMILES: O=C(C1=C(C)NC(C)=C(C(O)=O)C1C2=CC=CC(Cl)=C2Cl)OC
  • Shipping: Room temperature in continental US
  • Storage: Store at room temperature for 3 years; in solvent store at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-01667 Supplier Medchemexpress LLC Supplier No. HY330135G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clevidipine butyrate impurity 1 is an impurity of Clevidipine butyrate. It is for research use only and not intended for human use. The product is sold to researchers and not to patients.

  • Appearance: Off-white to light yellow solid
  • Formula: C16H15Cl2NO4
  • SMILES: O=C(C1=C(C)NC(C)=C(C(O)=O)C1C2=CC=CC(Cl)=C2Cl)OC
  • Shipping: Room temperature in continental US
  • Storage: Store at room temperature for 3 years; in solvent store at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.