Learn More
Selleck Chemical LLC Clevidipine Butyrate S2080-10mg
SDP
Catalog No. 503108979
Encompass_Preferred
Clevidipine Butyrate(Cleviprex) is a dihydropyridine calcium channel blocker uses as an agent for the reduction of blood pressure
Catalog No. 50-310-8979 Supplier Selleck Chemical LLC Supplier No. S208010MG
Clevidipine Butyrate(Cleviprex) is a dihydropyridine calcium channel blocker uses as an agent for the reduction of blood pressure