Learn More
MSC Campbell Clevis Hook, 5/8 in, Grade 70, 18,100 lb Capacity
Catalog No. 5000515553
Encompass
This clevis hook is designed for demanding applications, featuring a 5/8 in chain diameter and a Grade 70 rating. It offers a substantial work load limit, making it suitable for various industrial and rigging tasks.
  • 5/8 in chain diameter
  • Grade 70
  • 18,100 lb work load limit
  • 13/16 in inside diameter
  • 25/32 in pin diameter
  • 3/4 in hook throat
  • Alloy steel material
Catalog No. 50-005-15553 Supplier MSC Supplier No. 67782060
This clevis hook is designed for demanding applications, featuring a 5/8 in chain diameter and a Grade 70 rating. It offers a substantial work load limit, making it suitable for various industrial and rigging tasks.
  • 5/8 in chain diameter
  • Grade 70
  • 18,100 lb work load limit
  • 13/16 in inside diameter
  • 25/32 in pin diameter
  • 3/4 in hook throat
  • Alloy steel material