Learn More
Medchemexpress LLC Clevudine | 163252-36-6 | 99.84% | 260.22 | 100 MG
SDP
Catalog No. 5000352455
Encompass_Preferred
Clevudine (L-FMAU) is a nucleoside analog of the unnatural L-configuration, exhibiting potent anti-HBV activity with a long half-life and low toxicity. It acts as a non-competitive inhibitor by binding to the polymerase rather than being incorporated into the viral DNA. This compound is also effective against cowpox virus respiratory infection in mice and shows activity against Epstein-Barr virus.
  • Nucleoside analog with L-configuration
  • Potent anti-HBV activity
  • Long half-life and low toxicity
  • Non-competitive inhibitor; binds to polymerase
  • Effective against cowpox virus respiratory infection
  • Shows activity against Epstein-Barr virus
  • White to off-white solid appearance
Catalog No. 50-003-52455 Supplier Medchemexpress LLC Supplier No. HY13859100MG
Clevudine (L-FMAU) is a nucleoside analog of the unnatural L-configuration, exhibiting potent anti-HBV activity with a long half-life and low toxicity. It acts as a non-competitive inhibitor by binding to the polymerase rather than being incorporated into the viral DNA. This compound is also effective against cowpox virus respiratory infection in mice and shows activity against Epstein-Barr virus.
  • Nucleoside analog with L-configuration
  • Potent anti-HBV activity
  • Long half-life and low toxicity
  • Non-competitive inhibitor; binds to polymerase
  • Effective against cowpox virus respiratory infection
  • Shows activity against Epstein-Barr virus
  • White to off-white solid appearance