Learn More
Hach Company CLF10sc Total CL/SC4500 2Ch
Catalog No. 5000207365 Shop All Hach Company Products
Encompass
CLT10sc Total Chlorine Sensor-with SC4500 Controller-2 Channels-Grab SampleProviding the easy to integrate and upgrade SC4500 controller with the proven CL10sc sensor solution in one part number making ordering simple and fast For detailed technical specifications please visit the following part-numbers SC4500 LXV525 99A11501CLT10sc Sensor 8628900
Catalog No. 50-002-07365 Supplier Hach Company Supplier No. LXV525.97KTO029
Add to Cart
Add to Cart
Request a Quote
CLT10sc Total Chlorine Sensor-with SC4500 Controller-2 Channels-Grab SampleProviding the easy to integrate and upgrade SC4500 controller with the proven CL10sc sensor solution in one part number making ordering simple and fast For detailed technical specifications please visit the following part-numbers SC4500 LXV525 99A11501CLT10sc Sensor 8628900