Learn More
Rockland Immunochemicals Anti-Collagen I, alpha 1 telopeptide (Rabbit) Antibody - 600-401-D20
SDP
Supplier:  Rockland Immunochemicals 600401D20
Encompass
Anti-Collagen 1 alpha 1 telopeptide Antibody is suitable for use in ELISA, Western Blotting and IHC. The antibody works well for IHC on paraformaldehyde-fixed sections with a simple antigen-retrieval protocol (incubate slides for 20 minutes at 90º C in 10 mM sodium citrate (pH 6.0)/ 0.1 % Tween-20). Note that in paraffin sections of formaldehyde-fixed fibrotic mouse lung tissue, the antibody recognizes mature collagen I that has formed fibrils in the extracellular matrix. For Western blot with appropriately prepared samples expect a band of approximately 140 kDa in size corresponding to the telopeptide portion (C-terminus) of the collagen I alpha I polypeptide. Specific conditions for reactivity should be optimized by the end user.
Catalog No. NC0939456