Learn More
Aviva Systems Biology Corp CLGN Antibody - N-terminal region (ARP46436_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46436_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CLGN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CLGN. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53479