Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories CLIA Waived Fentanyl Test Cassette

Catalog No. 23111439
Change view
Click to view available options
Detectable Analytes:
Fentanyl (FEN)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-111-439 Fentanyl (FEN)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-439 Supplier Germaine™ Laboratories Supplier No. DFYA102C
This item is not returnable. View return policy

CLIA Waived Fentanyl Urine Test Cassette is a competitive binding, lateral flow immunochromatographic assay for qualitative detection of Fentanyl in human urine at the cutoff concentrations of 1 ng/mL.

  • Results in 5 minutes
  • 25 individually wrapped test cassettes per kit.
  • Approved for over the counter and professional use.

Specifications

CE Marker No
Clia Complexity Waived
Detectable Analytes Fentanyl (FEN)
Format Cassette
Sample Type Urine
Sensitivity 1 ng/mL
Type Fentanyl Test Cassette
Disposable Single-use
For Use With (Application) In Vitro Diagnostic Use
Analysis Time 5 mins.
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.