Learn More
Cliawaived Inc CLIAwaived, Inc. Drug Test Cup, 15 Panel, Fentanyl/Nor Fentanyl, FDA-Cleared
SDP
Catalog No. NC3980149
Encompass_Preferred
The INSTANT DRUG TEST CUP PLUS is a 15-panel, no-leak drug test cup that includes Fentanyl/Nor Fentanyl and adulterants. It provides results in 5 minutes and is FDA-cleared and CLIA-waived.
  • 15-panel drug test
  • No-leak cup design
  • Includes Fentanyl/Nor Fentanyl
  • Includes adulterants (OX/pH/SG), (GL/NI/CR)
  • Results in 5 minutes
  • FDA-cleared and CLIA-waived
  • Tests for AMP500, BAR300, BUP10, BZO300, COC150, EDDP300, FEN-NORFEN, MDMA500, MET500, MTD300, MOR300, OXY100, PCP25, TCA1000, THC50
Catalog No. NC3980149 Supplier Cliawaived Inc Supplier No. CLIAIDTC15BUPA
The INSTANT DRUG TEST CUP PLUS is a 15-panel, no-leak drug test cup that includes Fentanyl/Nor Fentanyl and adulterants. It provides results in 5 minutes and is FDA-cleared and CLIA-waived.
  • 15-panel drug test
  • No-leak cup design
  • Includes Fentanyl/Nor Fentanyl
  • Includes adulterants (OX/pH/SG), (GL/NI/CR)
  • Results in 5 minutes
  • FDA-cleared and CLIA-waived
  • Tests for AMP500, BAR300, BUP10, BZO300, COC150, EDDP300, FEN-NORFEN, MDMA500, MET500, MTD300, MOR300, OXY100, PCP25, TCA1000, THC50