Learn More
Aviva Systems Biology Corp CLIC1 Antibody - C-terminal region (ARP35044_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35044_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLIon Channel1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CLIC1. Alternate names: G6, NCC27ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55634