Learn More
Aviva Systems Biology Corp CLIC2 Antibody - middle region (ARP35047_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35047_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CLIon Channel2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CLIC2. Alternate names: CLIC2b, XAP121ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-55638