Learn More
Aviva Systems Biology Corp CLIC3 Antibody - N-terminal region (ARP35146_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35146_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLIC3. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-284-447