Learn More
Aviva Systems Biology Corp CLIC4 Antibody - N-terminal region (ARP35222_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35222_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CLIon Channel4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CLIC4. Alternate names: H1, huH1, p64H1, CLIC4L, MTCLICApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-55671