Learn More
Aviva Systems Biology Corp CLIC5 Antibody - middle region (ARP35262_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35262_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CLIon Channel5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human CLIC5. Alternate names: MST130, MSTP130ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55685