Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Click-iT™ EdU Colorimetric IHC Detection Kit

Catalog No. C10644
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10644 50 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10644 Supplier Invitrogen™ Supplier No. C10644
Only null left
Add to Cart
Edge
Add to Cart

Invitrogen™ Click-iT™ EdU Colorimetric IHC Detection Kit is a superior alternative to traditional methods for the colorimetric detection of proliferating cells within tissue sections.

  • Substrate: DAB
  • Substrate Type: HRP
  • The Click-iT™ EdU Colorimetric IHC (Immunohistochemistry) Detection Kit is a superior alternative to traditional methods for the colorimetric detection of proliferating cells within tissue sections. In this assay, 5-ethynyl-2'-deoxyuridine (EdU), a nucleoside analog of thymidine containing an alkyne moiety, is incorporated into newly synthesized DNA. When biotin containing an azide group is added, a brief click reaction attaches the azide to the alkyne moiety on the EdU molecule. Upon addition of streptavidin–peroxidase and a peroxidase substrate, an enzymatic reaction generates a signal that can be detected with light microscopy and stored for future analysis.
  • Optimized - enables colorimetric detection of proliferating cells in either tissue or cell samples
  • Efficient - no denaturation steps or harsh treatment required
  • Robust - improved preservation of cell morphology and the cellular environment offers content-rich results
  • Consistent - not dependent on variable antibody lots for detection
  • Simple - designed to work consistently, in less time than traditional methods
TRUSTED_SUSTAINABILITY

Specifications

Conjugate HRP
Detection Method Colorimetric
For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS™ XL Imaging System
Quantity 50 Slides
Packaging Slides
Content And Storage Store at 2°C to 8°C.
Shipping Condition Wet Ice
Substrate DAB
Substrate Type HRP
Product Line Click-iT
For Use With (Application) IHC (Immunohistochemistry)
Product Type IHC Detection Kit
Target Species All species
Show More Show Less
I have the Click-iT EdU Colorimetric IHC Detection Kit, but have decided that I want to use a fluorescent label but I don't want to buy another kit. What can I do?

You need to only substitute the peroxidase-conjugated streptavidin with a dye-conjugated streptavidin and not use the DAB. You would have to optimize the final concentration of the dye-conjugated streptavidin to use.

Can other peroxidase substrates be used with the Click-iT EdU Colorimetric IHC Detection Kit instead of the DAB?

Yes, but we have not optimized the amount of substrate to use. The labeling may be punctate and therefore an intense dark label such as DAB is recommended.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.