Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Click-iT™ sDIBO Alkynes for copper-free click chemistry
GREENER_CHOICE

Catalog No. C20022
Encompass_Preferred
Change view
Click to view available options
Label or Dye:
Alexa Fluor™ 488
Alexa Fluor™ 555
Alexa Fluor™ 594
Alexa Fluor™ 647
Biotin
SDP ester
Reactive Moiety:
DIBO Alkyne
SDP ester, sDIBO Alkyne
sDIBO Alkyne
Molecular Weight (g/mol):
∼1000
∼1050
∼1200
∼1400
∼1800
∼1900
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye Reactive Moiety Molecular Weight (g/mol)
C20022 Alexa Fluor™ 647 sDIBO Alkyne ∼1900
C10407 Alexa Fluor™ 594 DIBO Alkyne ∼1200
C20020 Alexa Fluor™ 488 sDIBO Alkyne ∼1400
C20021 Alexa Fluor™ 555 sDIBO Alkyne ∼1800
C20023 Biotin sDIBO Alkyne ∼1000
C20025 SDP ester SDP ester, sDIBO Alkyne ∼1050
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. C20022 Supplier Invitrogen™ Supplier No. C20022
Add to Cart
Edge
Add to Cart

Experience copper-free 'click chemistry' with Click-IT™ Alexa Fluor™, and sDIBO Alkyne reagents, which are ideal for surface labeling of live cells and minimize damage to enzymes and fluorescent proteins.

Achieve efficient labeling of azides with Click-It sDIBO Alkynes. Our Click-IT sDIBO alkynes enable the labeling of azido labeled targets without the need for copper catalysis and are available with a variety of Alexa Fluor or biotin labels, as well as amine or thiol reactive groups for creating your own conjugates. Macromolecules that have been azido-modified enzymatically, chemically, or metabolically can now be labeled easily, yielding soluble bioconjugates.

Click-iT sDIBO Alkyne labels react with azides via a copper-free Click chemistry reaction to form a stable triazole linkage with an Alexa Fluor, Biotin, or label of your choice. Copper-free click reactions using our sDIBO alkynes are ideal for surface labeling of live cells; they also minimize damage to fluorescent proteins like GFP or R-PE. For intracellular labeling, the nonspecific binding of DIBO alkynes creates a background signal, and a labeling strategy using picolyl azides is the preferred solution in these instances.

Click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background. Macromolecules that have been azide-modified enzymatically, chemically, or metabolically can be labeled more easily, yielding more soluble bioconjugates with improved biological labeling utility.

To make a concentrated stock solution from 2–5 mM, dissolve the dye-alkyne in high quality anhydrous DMSO. Store frozen, desiccated, and protected from light. Click-iT SDP Ester sDIBO alkyne is susceptible to hydrolysis, so prepare the stock solution fresh on the day of the experiment.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.5 mg
Kit Contents Contains 0.5 mg of Alexa Fluor 647 sDIBO Alkyne
Molecular Weight (g/mol) ∼1900
Filter Type Cy5
Content And Storage Store at ≤ -20°C. Desiccation recommended. Protect material from long-term exposure to light; may be exposed to light for short periods of time
Flow Cytometer Laser Lines Red
Labeling Method Click Chemistry
Shipping Condition Dry Ice
Reactive Moiety sDIBO Alkyne
Label or Dye Alexa Fluor™ 647
Color Far-red
Product Line Alexa Fluor, Click-iT
Emission 650 nm
Excitation 665 nm
Labeling Target Azide
Show More Show Less
What concentration and solvent do you recommend for making a stock solution of Click-iT sDIBO Alkynes for copper-free click chemistry?

For Click-iT sDIBO Alkynes for coper-free click chemistry (Cat. Nos. C10407, C20020, C20021, C20022, C20023, C20025), we recommend a 2-5 mM stock solution in DMSO.

How is SiteClick Alexa Fluor 647 sDIBO Alkyne different from Click-iT Alexa Fluor 647 sDIBO Alkyne?

Structurally, they are the same. The only difference is in the manner of packaging. SiteClick Alexa Fluor 647 sDIBO Alkyne is packaged for labeling 1 to 5 mg of antibody per the SiteClick labeling protocol with the actual amount being proprietary. Click-iT Alexa Fluor 647 sDIBO Alkyne is packaged as a 0.5 mg of reagent for use with any copper-free click reaction.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.