Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Click-On Inline Super Clean™ Gas Purifiers

Catalog No. 111016550 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Type:
Dual Trap (Hydrocarbon, Moisture)
Oxygen Trap
Triple Trap (Hydrocarbon, Moisture, Oxygen)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
11-101-6550 Dual Trap (Hydrocarbon, Moisture)
11-102-0688 Triple Trap (Hydrocarbon, Moisture, Oxygen)
11-101-6548 Oxygen Trap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-6550 Supplier MilliporeSigma™ Supelco™ Supplier No. 28866U
Add to Cart
Add to Cart

Simple installation and replacement of the Click-On Purifier eliminates the risk of damage to gas line connections caused by overtightening

  • Simply attach the connectors to the gas line, install the purifier, and hand tighten the connectors
  • Click-on Connectors are not included and must be purchased before initial use
TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 194 mm
Material Stainless Steel
Quantity 1 Ea.
Description For Fuel Gas
Type Dual Trap (Hydrocarbon, Moisture)
Diameter (Metric) Outer 32 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.