Learn More
MilliporeSigma™ Supelco™ Click-On Inline Super Clean™ Gas Purifiers
Shop All MilliporeSigma Supelco ProductsDescription
- Simply attach the connectors to the gas line, install the purifier, and hand tighten the connectors
- Click-on Connectors are not included and must be purchased before initial use
Specifications
Specifications
| Length (Metric) | 194 mm |
| Material | Stainless Steel |
| Quantity | 1 Ea. |
| Product Type | Triple Trap (Hydrocarbon, Moisture, Oxygen) |
| Diameter (Metric) Outer | 32 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.