Learn More
Carbosynth LLC. Clickable-TAT Alkyne (49-57), 1mg, MW: (M.W. 1432.76), Molecular formula: C59H113N31O11
SDP
Supplier:  Carbosynth LLC. TAT3733PI1MG
Encompass_Preferred
Synonyms: 5-hexynoyl-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-NH2, 5-hexynoyl-RKKRRQRRR-NH2, Application: Modified HIV TAT Sequence 49-57 for Click Chemistry, Salt Form:, Trifluoroacetate Form, Storage Temperature: -20°C, References: S. Brown and D. Graham, Tet Lett, 51, 5032 (2010).
Catalog No. 50-168-7184