Learn More
FUJIFILM Biosciences Clicks Medium EHAA 500ML
Catalog No. 502892160
Encompass_Preferred
Clicks Medium is a modification of Eagles Essential Medium with Hanks Salts (HMEM) and contains higher concentrations of essential amino acids, in addition to non-essential amino acids, sodium pyruvate, and nucleic acid precursors.With:1350 mg/L Sodium BicarbonateWithout:MercaptoethanolL-Glutamine - add 20mL/L (200mM)
Catalog No. 50-289-2160 Supplier FUJIFILM Biosciences Supplier No. 9195500ML
Clicks Medium is a modification of Eagles Essential Medium with Hanks Salts (HMEM) and contains higher concentrations of essential amino acids, in addition to non-essential amino acids, sodium pyruvate, and nucleic acid precursors.With:1350 mg/L Sodium BicarbonateWithout:MercaptoethanolL-Glutamine - add 20mL/L (200mM)