Learn More
TARGETMOL CHEMICALS INC Clidinium bromide 500MG
SDP
Catalog No. 502880345
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Clidinium bromide (Ro 2-3773) , a synthetic anticholinergic agent, has been shown in experimental and clinical researchers to have a pronounced antisecretory and antispasmodic effect on the gastrointestinal tract. It can inhibit muscarinic actions of acetylcholine at postganglionic parasympathetic neuroeffector sites. It is used for the treatment of peptic ulcer disease and also to help relieve cramps or stomach spasms or abdominal due to colicky abdominal diverticulitis, pain, and irritable bowel syndrome. Purity 99.36%
Catalog No. 50-288-0345 Supplier TARGETMOL CHEMICALS INC Supplier No. T0786500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Clidinium bromide (Ro 2-3773) , a synthetic anticholinergic agent, has been shown in experimental and clinical researchers to have a pronounced antisecretory and antispasmodic effect on the gastrointestinal tract. It can inhibit muscarinic actions of acetylcholine at postganglionic parasympathetic neuroeffector sites. It is used for the treatment of peptic ulcer disease and also to help relieve cramps or stomach spasms or abdominal due to colicky abdominal diverticulitis, pain, and irritable bowel syndrome. Purity 99.36%