Learn More
Cayman Chemical Clidinium (bromide), Purity: ≥98%, CAS: 3485-62-9, 1 g
SDP
Catalog No. 502964396
Encompass_Preferred
A gastrointestinal muscarinic receptor antagonist (Ki = 3 nM against [3H]quinuclidinyl benzilate binding in rat colonic enterocytes); a dose of 1 mg/kg slows intestinal transit and induces constipation in mice; induces serum hyperammonemia; decreases clonic, myoclonic, and tonic seizure thresholds in a murine pentylenetetrazole-induced epilepsy model
Catalog No. 50-296-4396 Supplier Cayman Chemical Supplier No. 214281 G
A gastrointestinal muscarinic receptor antagonist (Ki = 3 nM against [3H]quinuclidinyl benzilate binding in rat colonic enterocytes); a dose of 1 mg/kg slows intestinal transit and induces constipation in mice; induces serum hyperammonemia; decreases clonic, myoclonic, and tonic seizure thresholds in a murine pentylenetetrazole-induced epilepsy model