Learn More
Pharmacal Research Laboratories CLIDOX-S ACTIVATOR 1 GAL
SDP
Catalog No. NC0332382
Encompass
Clidox-S is a chlorine dioxide based sterilant . This product is Sporicidal, Tuberculocidal, Bactericidal, Virucidal and Fungicidal. Clidox-S can be mixed at 1:3:1 or 1:18:1 . This product kills both enveloped and non-enveloped viruses.
This item is not returnable. View return policy
Catalog No. NC0332382 Supplier Pharmacal Research Laboratories Supplier No. 95118F
Clidox-S is a chlorine dioxide based sterilant . This product is Sporicidal, Tuberculocidal, Bactericidal, Virucidal and Fungicidal. Clidox-S can be mixed at 1:3:1 or 1:18:1 . This product kills both enveloped and non-enveloped viruses.