Learn More
Pharmacal Research Laboratories CLIDOX-S ACTIVATOR 5GAL
SDP
Catalog No. NC9703273
Encompass
Clidox-S Activator, 5 gal, Components: Hydroxyacetic Acid, Conc.: 10% (approx.), Boiling Point: 112 deg.C, Specific Gravity: 1.03, pH: 2.0, Appearance: Clear Liquid, Pour Point: 10 deg.C, Volatility by Volume: 4.0%
This item is not returnable. View return policy
Catalog No. NC9703273 Supplier Pharmacal Research Laboratories Supplier No. 95125F
Clidox-S Activator, 5 gal, Components: Hydroxyacetic Acid, Conc.: 10% (approx.), Boiling Point: 112 deg.C, Specific Gravity: 1.03, pH: 2.0, Appearance: Clear Liquid, Pour Point: 10 deg.C, Volatility by Volume: 4.0%