Learn More
Pharmacal Research Laboratories Clidox -S Disinfectant 1 Gallon
SDP
Catalog No. NC9189926
Encompass
Clidox -S Disinfectant, Is a chlorine dioxide based disinfectant, Is tuberculocidal, bactericidal, virucidal and fungicidal, 1 gal.
This item is not returnable. View return policy
Catalog No. NC9189926 Supplier Pharmacal Research Laboratories Supplier No. 96118F
Clidox -S Disinfectant, Is a chlorine dioxide based disinfectant, Is tuberculocidal, bactericidal, virucidal and fungicidal, 1 gal.