Learn More
Pharmacal Research Laboratories CLIDOX-S BASE 4X1GAL
SDP
Catalog No. NC0089321
Encompass
Clidox-S is a chlorine dioxide based sterilant . This product is Sporicidal, Tuberculocidal, Bactericidal, Virucidal and Fungicidal. Clidox-S can be mixed at 1:3:1 or 1:18:1 . This product kills both enveloped and non-enveloped viruses. Clidox-S kills Mycobacterium bovis in 5 minutes at 20oC when used on precleaned surfaces. This product completely inactivates Minute Virus of Mice (Parvovirus) ATCC-VR-1346 when diluted 1:18:1 at 5 minutes at 21.5oC in the presence of 1% organic soil load. This product is virucidal (completely kills Herpes Simplex type 2 and Poliovirus type) when diluted at 1:18:1 at 3 minutes at 20oC on precleaned surfaces. 4x1 gallon.
This item is not returnable. View return policy
Catalog No. NC0089321 Supplier Pharmacal Research Laboratories Supplier No. 96120F
Clidox-S is a chlorine dioxide based sterilant . This product is Sporicidal, Tuberculocidal, Bactericidal, Virucidal and Fungicidal. Clidox-S can be mixed at 1:3:1 or 1:18:1 . This product kills both enveloped and non-enveloped viruses. Clidox-S kills Mycobacterium bovis in 5 minutes at 20oC when used on precleaned surfaces. This product completely inactivates Minute Virus of Mice (Parvovirus) ATCC-VR-1346 when diluted 1:18:1 at 5 minutes at 21.5oC in the presence of 1% organic soil load. This product is virucidal (completely kills Herpes Simplex type 2 and Poliovirus type) when diluted at 1:18:1 at 3 minutes at 20oC on precleaned surfaces. 4x1 gallon.