Learn More
Pharmacal Research Laboratories CLIDOX-S BASE 5 GALLON
SDP
Catalog No. NC9703274
Encompass
Clidox-S Base, 5 gal., Components: Sodium Chlorite, Conc.: 0.85% (approx.), Boiling Point: 100degreeC, Specific Gravity: 1.01, pH: 11.5, Appearance: Clear Liquid, Solubility in Water @ 1 Atm and 25degreeC: Complete
This item is not returnable. View return policy
Catalog No. NC9703274 Supplier Pharmacal Research Laboratories Supplier No. 96125F
Clidox-S Base, 5 gal., Components: Sodium Chlorite, Conc.: 0.85% (approx.), Boiling Point: 100degreeC, Specific Gravity: 1.01, pH: 11.5, Appearance: Clear Liquid, Solubility in Water @ 1 Atm and 25degreeC: Complete