Learn More
Medchemexpress LLC Climbazole-d4 | 1185117-79-6 | 99.9% | 1 MG
SDP
Catalog No. 5000412600
Encompass_Preferred
Climbazole-d4 is a deuterium-labeled version of Climbazole. Climbazole is recognized as a potent antifungal agent and also acts as a strong inducer of rat hepatic cytochrome P450. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules, primarily to serve as tracers for quantitative analysis during drug development, and this substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Is a deuterium-labeled compound.
  • The parent compound is a potent antifungal agent.
  • Is also a potent inducer of rat hepatic cytochrome P450.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques like NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12600 Supplier Medchemexpress LLC Supplier No. HYB1151S1MG
Climbazole-d4 is a deuterium-labeled version of Climbazole. Climbazole is recognized as a potent antifungal agent and also acts as a strong inducer of rat hepatic cytochrome P450. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules, primarily to serve as tracers for quantitative analysis during drug development, and this substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Is a deuterium-labeled compound.
  • The parent compound is a potent antifungal agent.
  • Is also a potent inducer of rat hepatic cytochrome P450.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques like NMR, GC-MS, or LC-MS.