Learn More
Cayman Chemical (±)-Climbazole-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1185117-79-6, 1 mg
SDP
Catalog No. 502940288
Encompass_Preferred
An internal standard for the quantification of climbazole by GC- or LC-MS
Catalog No. 50-294-0288 Supplier Cayman Chemical Supplier No. 276161 MG
An internal standard for the quantification of climbazole by GC- or LC-MS