Learn More
Medchemexpress LLC Climbazole Standard | 38083-17-9 | 97.8% | 250 MG
SDP
Catalog No. 5000413949
Encompass_Preferred
Climbazole (Standard) is the analytical standard for Climbazole. It is intended for research and analytical applications. Climbazole (BAY-e 6975) is a potent antifungal agent and also acts as a potent inducer of rat hepatic cytochrome P450.
  • Analytical standard for Climbazole
  • Intended for research and analytical applications
  • Potent antifungal agent
  • Potent inducer of rat hepatic cytochrome P450
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-13949 Supplier Medchemexpress LLC Supplier No. 250MGHYB1151R
Climbazole (Standard) is the analytical standard for Climbazole. It is intended for research and analytical applications. Climbazole (BAY-e 6975) is a potent antifungal agent and also acts as a potent inducer of rat hepatic cytochrome P450.
  • Analytical standard for Climbazole
  • Intended for research and analytical applications
  • Potent antifungal agent
  • Potent inducer of rat hepatic cytochrome P450
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS