Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Climbazole 98.0+%, TCI America™
SDP

Catalog No. C202525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C202525G 25 g
C2025500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C202525G Supplier TCI America Supplier No. C202525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38083-17-9
Molecular Formula C15H17ClN2O2
Molecular Weight (g/mol) 292.763
InChI Key OWEGWHBOCFMBLP-UHFFFAOYSA-N
Synonym 1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethyl-2-butanone
PubChem CID 37907
ChEBI CHEBI:83719
IUPAC Name 1-(4-chlorophenoxy)-1-imidazol-1-yl-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)C(N1C=CN=C1)OC2=CC=C(C=C2)Cl

Specifications

Melting Point 98°C
Color White
Quantity 25 g
Formula Weight 292.76
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Climbazole
RTECSNumber EL7085000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.