Learn More
Enzo Life Sciences CLIMP-63 monoclonal antibody (G1/296) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6690100
Encompass_Preferred
Clone: G1/296. Isotype: IgG2aκ. Immunogen: Human CLIMP-63 (cytoskeleton-linking membrane protein of 63kDa) from Caco-2 cells golgi membrane fraction. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Monkey. Applications: ELISA, Flow Cytometry, ICC, IHC (FS), IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.1% sodium azide.
Catalog No. 50-201-3196