Learn More
Apexbio Technology LLC Clinafloxacin CI96 AM1091 105956-97-6 250mg
SDP
Catalog No. 5000093930
Encompass_Preferred
Clinafloxacin CI96 AM1091 (CAS 105956-97-6) is a small-molecule inhibitor targeting bacterial DNA gyrase and topoisomerase IV It is designed to inhibit these type II topoisomerases thereby disrupting bacterial DNA replication and transcription Clinafloxacin CI96 AM1091 exerts its biological activity primarily through dual inhibition of DNA gyrase and topoisomerase IV In enzymatic assays against Staphylococcus aureus clinafloxacin demonstrates inhibitory activity with IC50 values of 0 92 g/mL for DNA gyrase and 1 62 g/mL for topoisomerase IV Based on these pharmacological properties clinafloxacin CI96 AM1091 holds research potential in bacteriology studies of bacterial resistance mechanisms and evaluation of drug-resistant bacterial strains
Catalog No. 50-000-93930 Supplier Apexbio Technology LLC Supplier No. A8400250
Clinafloxacin CI96 AM1091 (CAS 105956-97-6) is a small-molecule inhibitor targeting bacterial DNA gyrase and topoisomerase IV It is designed to inhibit these type II topoisomerases thereby disrupting bacterial DNA replication and transcription Clinafloxacin CI96 AM1091 exerts its biological activity primarily through dual inhibition of DNA gyrase and topoisomerase IV In enzymatic assays against Staphylococcus aureus clinafloxacin demonstrates inhibitory activity with IC50 values of 0 92 g/mL for DNA gyrase and 1 62 g/mL for topoisomerase IV Based on these pharmacological properties clinafloxacin CI96 AM1091 holds research potential in bacteriology studies of bacterial resistance mechanisms and evaluation of drug-resistant bacterial strains