Learn More
Medchemexpress LLC Clinafloxacin (hydrochloride) | 105956-99-8 | 99.6% | 5 MG
SDP
Catalog No. 5000405487
Encompass_Preferred
Clinafloxacin hydrochloride (AM 1091 hydrochloride) is a potent, broad-spectrum fluoroquinolone antibiotic with inhibitory activity against gram-positive, gram-negative bacteria, and anaerobic pathogens. It targets DNA gyrase and topoisomerase IV, making it suitable for laboratory research.
  • Potent and broad-spectrum fluoroquinolone antibiotic
  • Active against gram-positive, gram-negative, and anaerobic pathogens
  • Targets DNA gyrase and topoisomerase IV
  • For research use only
  • Soluble in DMSO
Catalog No. 50-004-05487 Supplier Medchemexpress LLC Supplier No. HYB0536A5MG
Clinafloxacin hydrochloride (AM 1091 hydrochloride) is a potent, broad-spectrum fluoroquinolone antibiotic with inhibitory activity against gram-positive, gram-negative bacteria, and anaerobic pathogens. It targets DNA gyrase and topoisomerase IV, making it suitable for laboratory research.
  • Potent and broad-spectrum fluoroquinolone antibiotic
  • Active against gram-positive, gram-negative, and anaerobic pathogens
  • Targets DNA gyrase and topoisomerase IV
  • For research use only
  • Soluble in DMSO