Learn More
Medchemexpress LLC Clinafloxacin hydrochloride | 105956-99-8 | MFCD03840545 | 10 MG
SDP
Catalog No. 5000299026
Encompass_Preferred
Clinafloxacin hydrochloride is the hydrochloride salt of clinafloxacin, a potent broad-spectrum fluoroquinolone antibiotic used for research. It exhibits activity against Gram-positive and Gram-negative bacteria and is supplied as a research reagent for antibacterial studies and assay development.
  • Hydrochloride salt form for improved solubility.
  • Broad-spectrum fluoroquinolone antibacterial activity.
  • Suitable for in vitro antibacterial research and assay development.
  • Molecular weight 402.25 g/mol; CAS 105956-99-8.
  • Available in small research pack sizes, e.g. 10 mg.
Catalog No. 50-002-99026 Supplier Medchemexpress LLC Supplier No. HYB0536A10MG
Clinafloxacin hydrochloride is the hydrochloride salt of clinafloxacin, a potent broad-spectrum fluoroquinolone antibiotic used for research. It exhibits activity against Gram-positive and Gram-negative bacteria and is supplied as a research reagent for antibacterial studies and assay development.
  • Hydrochloride salt form for improved solubility.
  • Broad-spectrum fluoroquinolone antibacterial activity.
  • Suitable for in vitro antibacterial research and assay development.
  • Molecular weight 402.25 g/mol; CAS 105956-99-8.
  • Available in small research pack sizes, e.g. 10 mg.