Learn More
Cayman Chemical Clinafloxacin (hydrochloride), Purity: ≥95%, CAS: 105956-99-8, 100 mg
SDP
Catalog No. 502965045
Encompass_Preferred
A fluoroquinolone antibiotic that displays broad spectrum antibacterial activity by inhibiting the bacterial regulatory enzyme DNA gyrase (IC50 = 0.92 µg/ml) as well as topoisomerase IV (IC50 = 1.62 µg/ml)
Catalog No. 50-296-5045 Supplier Cayman Chemical Supplier No. 16923100 MG
A fluoroquinolone antibiotic that displays broad spectrum antibacterial activity by inhibiting the bacterial regulatory enzyme DNA gyrase (IC50 = 0.92 µg/ml) as well as topoisomerase IV (IC50 = 1.62 µg/ml)