Learn More
Selleck Chemical LLC Clindamycin S2830-1g
SDP
Catalog No. 503112879
Encompass_Preferred
Clindamycin inhibits protein synthesis by acting on the 50S ribosomal used for the treatment of bacterial infections
Catalog No. 50-311-2879 Supplier Selleck Chemical LLC Supplier No. S28301G
Clindamycin inhibits protein synthesis by acting on the 50S ribosomal used for the treatment of bacterial infections