Learn More
Apexbio Technology LLC Clindamycin 18323-44-9 100mg
SDP
Catalog No. 5000094679
Encompass_Preferred
Clindamycin (CAS 18323-44-9) is a small-molecule inhibitor targeting the bacterial 50S ribosomal subunit It is designed to disrupt peptide bond formation during protein synthesis thereby inhibiting bacterial proliferation Clindamycin exerts its biological activity primarily through binding to the 50S ribosomal subunit which interrupts peptide elongation and protein assembly In biochemical research Clindamycin demonstrates antimicrobial inhibition with reported IC50 values typically in the low micromolar range depending on bacterial strains and experimental conditions Based on these pharmacological properties Clindamycin holds research potential in microbiological and pharmacological studies for evaluating antimicrobial susceptibility bacterial ribosomal functions and mechanisms of antibiotic resistance
Catalog No. 50-000-94679 Supplier Apexbio Technology LLC Supplier No. B1707100
Clindamycin (CAS 18323-44-9) is a small-molecule inhibitor targeting the bacterial 50S ribosomal subunit It is designed to disrupt peptide bond formation during protein synthesis thereby inhibiting bacterial proliferation Clindamycin exerts its biological activity primarily through binding to the 50S ribosomal subunit which interrupts peptide elongation and protein assembly In biochemical research Clindamycin demonstrates antimicrobial inhibition with reported IC50 values typically in the low micromolar range depending on bacterial strains and experimental conditions Based on these pharmacological properties Clindamycin holds research potential in microbiological and pharmacological studies for evaluating antimicrobial susceptibility bacterial ribosomal functions and mechanisms of antibiotic resistance