Learn More
Medchemexpress LLC Clindamycin | 18323-44-9 | 99.9% | 1 ML
SDP
Catalog No. 5000413702
Encompass_Preferred
Clindamycin is an orally active and broad-spectrum bacteriostatic lincosamide antibiotic. It can inhibit bacterial protein synthesis and suppress the expression of virulence factors in Staphylococcus aureus at sub-inhibitory concentrations. Clindamycin resistance results from enzymatic methylation of the antibiotic binding site in the 50S ribosomal subunit (23S rRNA). It also decreases the production of panton-valentine leucocidin, toxic-shock-staphylococcal toxin, or alpha-haemolysin. Clindamycin can be used for researching malaria.
  • Inhibits bacterial protein synthesis.
  • Suppresses expression of virulence factors in Staphylococcus aureus.
  • Decreases the production of panton-valentine leucocidin, toxic-shock-staphylococcal toxin, or alpha-haemolysin.
  • Used for researching malaria.
Catalog No. 50-004-13702 Supplier Medchemexpress LLC Supplier No. HYB145510MM/1ML
Clindamycin is an orally active and broad-spectrum bacteriostatic lincosamide antibiotic. It can inhibit bacterial protein synthesis and suppress the expression of virulence factors in Staphylococcus aureus at sub-inhibitory concentrations. Clindamycin resistance results from enzymatic methylation of the antibiotic binding site in the 50S ribosomal subunit (23S rRNA). It also decreases the production of panton-valentine leucocidin, toxic-shock-staphylococcal toxin, or alpha-haemolysin. Clindamycin can be used for researching malaria.
  • Inhibits bacterial protein synthesis.
  • Suppresses expression of virulence factors in Staphylococcus aureus.
  • Decreases the production of panton-valentine leucocidin, toxic-shock-staphylococcal toxin, or alpha-haemolysin.
  • Used for researching malaria.