Learn More
Medchemexpress LLC Clindamycin | 18323-44-9 | 99.9% | 5 MG
SDP
Catalog No. 5000413427
Encompass_Preferred
Clindamycin is an orally active, broad-spectrum bacteriostatic lincosamide antibiotic. It inhibits bacterial protein synthesis and can suppress the expression of virulence factors in Staphylococcus aureus at sub-inhibitory concentrations (sub-MICs). Resistance to Clindamycin occurs due to enzymatic methylation of the antibiotic binding site in the 50S ribosomal subunit (23S rRNA). It reduces the production of Panton-Valentine leucocidin (PVL), toxic-shock-staphylococcal toxin (TSST-1), or alpha-haemolysin (Hla). Clindamycin is also used for researching malaria.
  • Orally active and broad-spectrum bacteriostatic lincosamide antibiotic.
  • Inhibits bacterial protein synthesis.
  • Suppresses virulence factors in Staphylococcus aureus.
  • Decreases production of PVL, TSST-1, or Hla.
  • Can be used for researching malaria.
  • For research use only.
Catalog No. 50-004-13427 Supplier Medchemexpress LLC Supplier No. HYB14555MG
Clindamycin is an orally active, broad-spectrum bacteriostatic lincosamide antibiotic. It inhibits bacterial protein synthesis and can suppress the expression of virulence factors in Staphylococcus aureus at sub-inhibitory concentrations (sub-MICs). Resistance to Clindamycin occurs due to enzymatic methylation of the antibiotic binding site in the 50S ribosomal subunit (23S rRNA). It reduces the production of Panton-Valentine leucocidin (PVL), toxic-shock-staphylococcal toxin (TSST-1), or alpha-haemolysin (Hla). Clindamycin is also used for researching malaria.
  • Orally active and broad-spectrum bacteriostatic lincosamide antibiotic.
  • Inhibits bacterial protein synthesis.
  • Suppresses virulence factors in Staphylococcus aureus.
  • Decreases production of PVL, TSST-1, or Hla.
  • Can be used for researching malaria.
  • For research use only.