Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clindamycin | 18323-44-9 | MFCD00072005 | 10 MG
SDP

Catalog No. 5000297813
Encompass_Preferred

Clindamycin is an orally active, broad-spectrum bacteriostatic lincosamide antibiotic used in research to inhibit bacterial protein synthesis and to study suppression of virulence factors in Staphylococcus aureus and Plasmodium models.

  • Broad-spectrum lincosamide antibiotic for research use.
  • Inhibits bacterial protein synthesis.
  • Suppresses virulence factor expression in Staphylococcus aureus at subinhibitory concentrations.
  • Reported activity in Plasmodium research.
  • Solubility: DMSO 125 mg/mL; H2O 2 mg/mL with warming and ultrasonic treatment.
  • Storage: -20°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Molecular weight 424.98; molecular formula C18H33ClN2O5S.

Catalog No. 50-002-97813 Supplier Medchemexpress LLC Supplier No. HYB145510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clindamycin is an orally active, broad-spectrum bacteriostatic lincosamide antibiotic used in research to inhibit bacterial protein synthesis and to study suppression of virulence factors in Staphylococcus aureus and Plasmodium models.

  • Broad-spectrum lincosamide antibiotic for research use.
  • Inhibits bacterial protein synthesis.
  • Suppresses virulence factor expression in Staphylococcus aureus at subinhibitory concentrations.
  • Reported activity in Plasmodium research.
  • Solubility: DMSO 125 mg/mL; H2O 2 mg/mL with warming and ultrasonic treatment.
  • Storage: -20°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Molecular weight 424.98; molecular formula C18H33ClN2O5S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.