Learn More
Selleck Chemical LLC Clindamycin alcoholate S5420-25mg
SDP
Catalog No. 503109953
Encompass_Preferred
Clindamycin alcoholate is a crystallizable compound of Clindamycin with alcohol Clindamycin is a semisynthetic lincosamide antibiotic that inhibits bacterial protein synthesis by binding to bacterial 50S ribosomal subunits
Catalog No. 50-310-9953 Supplier Selleck Chemical LLC Supplier No. S542025MG
Clindamycin alcoholate is a crystallizable compound of Clindamycin with alcohol Clindamycin is a semisynthetic lincosamide antibiotic that inhibits bacterial protein synthesis by binding to bacterial 50S ribosomal subunits