Learn More
Alkali Scientific Clindamycin HCl, 1 G
SDP
Catalog No. 5000660734
Encompass_Preferred
Clindamycin HCl is a lincosamide antibiotic used in microbiological and biochemical research for its ability to inhibit bacterial protein synthesis. It binds to the 50S ribosomal subunit, preventing peptide chain elongation and effectively suppressing bacterial growth. Clindamycin is particularly active against Gram-positive bacteria and certain anaerobic organisms, making it useful in selective culture applications. In laboratory settings, it is commonly used in antimicrobial studies, cell culture contamination control, and research involving bacterial resistance mechanisms. The compound is suitable for in vitro experiments and supports studies focused on translation inhibition, bacterial metabolism, and antibiotic efficacy testing.
Catalog No. 50-006-60734 Supplier Alkali Scientific Supplier No. CLI001CP
Clindamycin HCl is a lincosamide antibiotic used in microbiological and biochemical research for its ability to inhibit bacterial protein synthesis. It binds to the 50S ribosomal subunit, preventing peptide chain elongation and effectively suppressing bacterial growth. Clindamycin is particularly active against Gram-positive bacteria and certain anaerobic organisms, making it useful in selective culture applications. In laboratory settings, it is commonly used in antimicrobial studies, cell culture contamination control, and research involving bacterial resistance mechanisms. The compound is suitable for in vitro experiments and supports studies focused on translation inhibition, bacterial metabolism, and antibiotic efficacy testing.